3,5-Di-tert-butylbenzoesäure ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

L12386.03 L12386.06
L12386.03EA 19.9 EUR
L12386.03 L12386.06
3,5-Di-tert-butylbenzoesäure ≥99%
3,5-Di-tert-butylbenzoesäure

Purity: 99% CAS No.: 16225-26-6 Molecular Formula: C15H22O2 Formula Weight: 234.34 Melting Point: 160-162° Beilstein Reference: 981248

Formel: C₁₅H₂₂O₂
Schmelzpunkt: 160…162 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00082727
CAS-Nummer: 16225-26-6
EINECS: 240-350-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR