Lithium acetylide ethylenediamine complex 90%, tech.

Lieferant: Thermo Fisher Scientific
Gefahr

L12401.22
L12401.22EA 250 EUR
L12401.22
Lithium acetylide ethylenediamine complex 90%, tech.
Lithium acetylide ethylenediamine complex

Purity: 90% CAS No.: 6867-30-7 Molecular Formula: C4H9LiN2 Formula Weight: 92.07 Odour: Amine-like Melting Point: ca 76° dec. Beilstein Reference: 3593824 UN No.: UN3131 MDL No.: MFCD00013183

Formel: C₄H₉LiN₂
Schmelzpunkt: ∼ 76 °C (Zersetzung)
Dichte: 1,04
Lagertemperatur: Raumtemperatur
MDL: MFCD00013183
CAS-Nummer: 6867-30-7
EINECS: 229-967-9
UN: 3131
ADR: 4.3,I

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR