4'-Methyl-2,2,2-trifluoracetophenon ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2,2,2-Trifluor-1-(4-methylphenyl)-1-ethanon

L12424.03 L12424.06
L12424.03EA 64.4 EUR
L12424.03 L12424.06
4'-Methyl-2,2,2-trifluoracetophenon ≥97%
4'-Methyl-2,2,2-trifluoracetophenon

Purity: 97% CAS No.: 394-59-2 Molecular Formula: C9H7F3O Formula Weight: 188.15 Refractive Index: 1.4700 Boiling Point: 70-72°/15mm MDL No.: MFCD00052338

Formel: C₉H₇F₃O
Molecular Weight: 188.15 g/mol
Siedepunkt: 70…72 °C (15 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD00052338
CAS-Nummer: 394-59-2

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR