4-(Trifluormethyl)phenylhydrazin 95%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
L12436.03
L12436.06
L12436.03EA
35.8
EUR
L12436.03
L12436.06
4-(Trifluormethyl)phenylhydrazin 95%
4-(Trifluormethyl)phenylhydrazin
Purity: 95%
CAS No.: 368-90-1
Molecular Formula: C7H7F3N2
Formula Weight: 176.14
Storage Temperature: 2°C to 8°C
Melting Point: 60-64°
Boiling Point: 119-121°/17mm
Beilstein Reference: 743759
MDL No.: MFCD00042508
Formel:
C₇H₇F₃N₂ Siedepunkt: 119…121 °C (17 mmHg) Schmelzpunkt: 60…64 °C |
MDL:
MFCD00042508 CAS-Nummer: 368-90-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...