2-Hydroxythiophenol 97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 2-Hydroxybenzolthiol, o-Hydroxythiophenol, 2-Hydroxyphenylmercaptan, 1-Hydroxy-2-mercaptobenzol, 2-Mercaptophenol, 2-Hydroxybenzol-1-thiol

L12519.06
L12519.06EA 196 EUR
L12519.06
2-Hydroxythiophenol 97%
2-Hydroxythiophenol

Purity: 97% CAS No.: 1121-24-0 Molecular Formula: C6H6OS Formula Weight: 126.18 Flash Point: 97°(206°F) Refractive Index: 1.6140 Odour: Unpleasant Melting Point: 19° Boiling Point: 217° Beilstein Reference: 1280884 UN No.: 2922 MDL No.: MFCD00040447

Formel: C₆H₆OS
Molecular Weight: 126.18 g/mol
Siedepunkt: 217 °C
Schmelzpunkt: 19 °C
Dichte: 1.220
Flammpunkt: 97 °C (206 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00040447
CAS-Nummer: 1121-24-0
EINECS: 214-326-8
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR