N,N-Bis(trifluormethansulfonyl)anilin ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1,1,1-Trifluoro-N-phenyl-N-((trifluoromethyl)sulphonyl)methanesulphonamide, N-Phenylbis(trifluoromethanesulphonimide)

L12531.06 L12531.14
L12531.06EA 73.5 EUR
L12531.06 L12531.14
N,N-Bis(trifluormethansulfonyl)anilin ≥99%
N,N-Bis(trifluormethansulfonyl)anilin

Purity: 99% CAS No.: 37595-74-7 Molecular Formula: (CF3SO2)2NC6H5 Formula Weight: 357.25 Melting Point: 100-102° Beilstein Reference: 1269141

Formel: C₈H₅F₆NO₄S₂
Schmelzpunkt: 100…102 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00000404
CAS-Nummer: 37595-74-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR