N,N-Bis(trifluormethansulfonyl)anilin ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1,1,1-Trifluoro-N-phenyl-N-((trifluoromethyl)sulphonyl)methanesulphonamide, N-Phenylbis(trifluoromethanesulphonimide)
L12531.06
L12531.14
L12531.06EA
73.5
EUR
L12531.06
L12531.14
N,N-Bis(trifluormethansulfonyl)anilin ≥99%
N,N-Bis(trifluormethansulfonyl)anilin
Purity: 99%
CAS No.: 37595-74-7
Molecular Formula: (CF3SO2)2NC6H5
Formula Weight: 357.25
Melting Point: 100-102°
Beilstein Reference: 1269141
Formel:
C₈H₅F₆NO₄S₂ Schmelzpunkt: 100…102 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000404 CAS-Nummer: 37595-74-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...