trans-2-Octen ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 97%
CAS No.: 13389-42-9
Molecular Formula: C8H16
Formula Weight: 112.22
Flash Point: 14°(57°F)
Refractive Index: 1.4130
Melting Point: -89 to -86°
Boiling Point: 122-123°
Beilstein Reference: 1719497
UN No.: 3295
MDL No.: MFCD00009532
Formel:
C₈H₁₆ Siedepunkt: 122…123 °C Schmelzpunkt: –89 to –86 °C Dichte: 0,718 Flammpunkt: 14 °C (57 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009532 CAS-Nummer: 13389-42-9 EINECS: 236-463-2 UN: 3295 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...