Methyl-4-brombutyrat ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Methyl 4-bromobutanoate
L12636.09
L12636.09EA
36.4
EUR
L12636.09
Methyl-4-brombutyrat ≥97%
Methyl-4-brombutyrat
Purity: 97%
CAS No.: 4897-84-1
Molecular Formula: Br(CH2)3CO2CH3
Formula Weight: 181.04
Refractive Index: 1.4620
Boiling Point: 186-187°
MDL No.: MFCD00041482
Formel:
C₅H₉BrO₂ Molecular Weight: 181,03 g/mol Siedepunkt: 186…187 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041482 CAS-Nummer: 4897-84-1 EINECS: 225-523-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...