Triethylcitrat ≥99%
Lieferant: Thermo Fisher Scientific
Purity: 99%
CAS No.: 77-93-0
Molecular Formula: C12H20O7
Formula Weight: 276.29
Flash Point: 176°(348°F)
Refractive Index: 1.4420
Melting Point: -55°
Boiling Point: 126-127°/1mm
Beilstein Reference: 1801199
Merck Reference: 14,2326
MDL No.: MFCD00009201
Formel:
C₁₂H₂₀O₇ Siedepunkt: 126…127 °C (1 mmHg) Schmelzpunkt: –55 °C Dichte: 1,137 Flammpunkt: 176 °C (348 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009201 CAS-Nummer: 77-93-0 EINECS: 201-070-7 Merck Index: 14,2326 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...