Triethylcitrat ≥99%

Lieferant: Thermo Fisher Scientific

L12639.30 L12639.14 L12639.0B
L12639.30EA 31.8 EUR
L12639.30 L12639.14 L12639.0B
Triethylcitrat ≥99%
Triethylcitrat

Purity: 99% CAS No.: 77-93-0 Molecular Formula: C12H20O7 Formula Weight: 276.29 Flash Point: 176°(348°F) Refractive Index: 1.4420 Melting Point: -55° Boiling Point: 126-127°/1mm Beilstein Reference: 1801199 Merck Reference: 14,2326 MDL No.: MFCD00009201

Formel: C₁₂H₂₀O₇
Siedepunkt: 126…127 °C (1 mmHg)
Schmelzpunkt: –55 °C
Dichte: 1,137
Flammpunkt: 176 °C (348 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009201
CAS-Nummer: 77-93-0
EINECS: 201-070-7
Merck Index: 14,2326

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR