Dibutylestersulfat 95%

Lieferant: Thermo Fisher Scientific
Achtung

L12813.09 L12813.18
L12813.09EA 54.2 EUR
L12813.09 L12813.18
Dibutylestersulfat 95%
Dibutylestersulfat

Purity: 95% CAS No.: 625-22-9 Molecular Formula: C8H18O4S Formula Weight: 210.29 Refractive Index: 1.4205 Boiling Point: 115-116°/6mm MDL No.: MFCD00059420

Formel: C₈H₁₈O₄S
Siedepunkt: 115…116 °C (6 mmHg)
Dichte: 1,060
Lagertemperatur: Raumtemperatur
MDL: MFCD00059420
CAS-Nummer: 625-22-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR