5-Chlorobenzo[b]thiophene-3-acetic acid 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L12836.03
L12836.MD
L12836.03EA
94
EUR
L12836.03
L12836.MD
5-Chlorobenzo[b]thiophene-3-acetic acid 97%
5-Chlorobenzo[b]thiophene-3-acetic acid
Purity: 97%
CAS No.: 17266-30-7
Molecular Formula: C10H7ClO2S
Formula Weight: 226.68
Melting Point: 152-155°
Beilstein Reference: 1244795
MDL No.: MFCD00052308
Formel:
C₁₀H₇ClO₂S Schmelzpunkt: 152…155 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052308 CAS-Nummer: 17266-30-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...