3,6-Dichloranthranilsäure 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Amino-3,6-dichlorobenzoic acid

L12850.06
L12850.06EA 85.5 EUR
L12850.06
3,6-Dichloranthranilsäure 95%
3,6-Dichloranthranilsäure

Purity: 95% CAS No.: 3032-32-4 Molecular Formula: C7H5Cl2NO2 Formula Weight: 206.03 Melting Point: 150-154° Beilstein Reference: 3266879

Formel: C₇H₅Cl₂NO₂
Molecular Weight: 206,03 g/mol
Schmelzpunkt: 150…154 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052390
CAS-Nummer: 3032-32-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR