4,4'-Bismaleimidodiphenylmethane 95%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
1,1'-(Methylenebis(4,1-phenylene))bis(1H-pyrrole-2,5-dione), 4,4'-Methylenebis(N-phenylmaleimide)
L12862.14
L12862.14EA
51.4
EUR
L12862.14
4,4'-Bismaleimidodiphenylmethane 95%
4,4'-Bismaleimidodiphenylmethane
Purity: 95%
CAS No.: 13676-54-5
Molecular Formula: C21H14N2O4
Formula Weight: 358.36
Melting Point: 156-158°
Beilstein Reference: 333986
UN No.: UN2811
MDL No.: MFCD00005507
Formel:
C₂₁H₁₄N₂O₄ Molecular Weight: 358,35 g/mol Schmelzpunkt: 156…158 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005507 CAS-Nummer: 13676-54-5 EINECS: 237-163-4 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...