1,2-Ethandithiol ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
L12865.14
L12865.22
L12865.36
L12865.14EA
26.5
EUR
L12865.14
L12865.22
L12865.36
1,2-Ethandithiol ≥98%
1,2-Ethandithiol
Purity: 98+%
CAS No.: 540-63-6
Molecular Formula: C2H6S2
Formula Weight: 94.20
Flash Point: 45°(113°F)
Refractive Index: 1.5580
Odour: Stench
Melting Point: -41°
Boiling Point: 144-146°
Beilstein Reference: 505946
Merck Reference: 14,3725
UN No.: 3071
MDL No.: MFCD00004892
Formel:
C₂H₆S₂ Siedepunkt: 144…146 °C Schmelzpunkt: –41 °C Dichte: 1,123 Flammpunkt: 45 °C (113 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004892 CAS-Nummer: 540-63-6 EINECS: 208-752-3 UN: 3071 ADR: 6.1,II Merck Index: 14,3725 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...