6-Brom-1-hexen 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 95%
CAS No.: 2695-47-8
Molecular Formula: C6H11Br
Formula Weight: 163.06
Flash Point: 54°(129°F)
Refractive Index: 1.4652
Odour: Characteristic
Boiling Point: 147°
Beilstein Reference: 506203
UN No.: 1993
MDL No.: MFCD00000269
Formel:
C₆H₁₁Br Siedepunkt: 147 °C Dichte: 1,217 Flammpunkt: 54 °C (129 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000269 CAS-Nummer: 2695-47-8 EINECS: 220-267-9 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...