2-Hydroxy-4-methoxy-5-sulfobenzophenon Hydrat tech. 85% may cont. up to 10% 2-propanol, technische Qualität

Lieferant: Thermo Fisher Scientific
Achtung

L12912.09 L12912.18
L12912.09EA 49.5 EUR
L12912.09 L12912.18
2-Hydroxy-4-methoxy-5-sulfobenzophenon Hydrat tech. 85% may cont. up to 10% 2-propanol, technische Qualität
2-Hydroxy-4-methoxy-5-sulfobenzophenon Hydrat

Purity: 85% CAS No.: 4065-45-6 Molecular Formula: C14H12O6S.xH2O Formula Weight: 308.31(anhy) Melting Point: 108-112° Beilstein Reference: 2889165 Merck Reference: 14,8983 MDL No.: MFCD00977409

Formel: C₁₄H₁₂O₆S·H₂O
Schmelzpunkt: 108…112 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00024962
CAS-Nummer: 4065-45-6
EINECS: 223-772-2
Merck Index: 14,8983

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR