3,9-Bis(1,1-dimethyl-2-hydroxyethyl)-2,4,8,10-tetraoxaspiro[5.5]undecane 97%

Lieferant: Thermo Scientific

Synonyme: 2,2'-(2,4,8,10-Tetraoxaspiro[5.5]undecane-3,9-diyl)bis(2-methylpropan-1-ol), Bis(hydroxypivalaldehyde) Pentaerythritolacetal Cyclic Acetal

L12958.22
L12958.22EA 66 EUR
L12958.22
3,9-Bis(1,1-dimethyl-2-hydroxyethyl)-2,4,8,10-tetraoxaspiro[5.5]undecane 97%
3,9-Bis(1,1-dimethyl-2-hydroxyethyl)-2,4,8,10-tetraoxaspiro[5.5]undecane

Purity: 97% CAS No.: 1455-42-1 Molecular Formula: C15H28O6 Formula Weight: 304.39 Melting Point: 197-200° Beilstein Reference: 1574570 MDL No.: MFCD00059794

Formel: C₁₅H₂₈O₆
Molecular Weight: 304,38 g/mol
Schmelzpunkt: 197…200 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00059794
CAS-Nummer: 1455-42-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR