Natriumcarbonat ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: wasserfreie Soda

L13098.36 L13098.0E L13098.0I
L13098.36EA 27.2 EUR
L13098.36 L13098.0E L13098.0I
Natriumcarbonat ≥98%
Natriumcarbonat

Purity: 98% CAS No.: 497-19-8 Molecular Formula: Na2CO3 Formula Weight: 105.99 Refractive Index: 1.535 Odour: Odourless pH: 11,5 Beilstein Reference: 4154566 Merck Reference: 14,8596 MDL No.: MFCD00003494

Formel: Na₂CO₃
Molecular Weight: 105.99 g/mol
Siedepunkt: 1600
Schmelzpunkt: 854
Dichte: 2.53
Lagertemperatur: Raumtemperatur
MDL: MFCD00003494
CAS-Nummer: 497-19-8
EINECS: 207-838-8
REACH: 01-2119485498-19
Merck Index: 14,8596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR