2,6-Dibrom-4-nitrotoluol 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1,3-Dibromo-2-methyl-5-nitrobenzene
L13102.06
L13102.14
L13102.06EA
54
EUR
L13102.06
L13102.14
2,6-Dibrom-4-nitrotoluol 95%
2,6-Dibrom-4-nitrotoluol
Purity: 95%
CAS No.: 110127-07-6
Molecular Formula: C7H5Br2NO2
Formula Weight: 294.94
Melting Point: 54-57°
Beilstein Reference: 3259701
Formel:
C₇H₅Br₂NO₂ Schmelzpunkt: 54…57 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00151807 CAS-Nummer: 110127-07-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...