4-Nitro-m-toluidin ≥99%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 3-Methyl-4-nitroaniline, 4-Nitro-3-methylaniline, 5-Amino-2-nitrotoluene

L13112.06
L13112.06EA 74.9 EUR
L13112.06
4-Nitro-m-toluidin ≥99%
4-Nitro-m-toluidin

Purity: 99% CAS No.: 611-05-2 Molecular Formula: C7H8N2O2 Formula Weight: 152.15 Odour: Characteristic Melting Point: 133-137° Beilstein Reference: 2209112 UN No.: 2660 MDL No.: MFCD00091833

Formel: C₇H₈N₂O₂
Molecular Weight: 152,15 g/mol
Schmelzpunkt: 133…137 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00091833
CAS-Nummer: 611-05-2
EINECS: 210-247-8
UN: 2660
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR