4-Hydroxy-2-(trifluoromethyl)pyrimidine 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-(Trifluoromethyl)pyrimidin-4-ol

L13118.03
L13118.03EA 55.4 EUR
L13118.03
4-Hydroxy-2-(trifluoromethyl)pyrimidine 97%
4-Hydroxy-2-(trifluoromethyl)pyrimidine

Purity: 97% CAS No.: 1546-80-1 Molecular Formula: C5H3F3N2O Formula Weight: 164.09 Melting Point: 168-171° Beilstein Reference: 778887 MDL No.: MFCD00151833

Formel: C₅H₃F₃N₂O
Molecular Weight: 164,09 g/mol
Schmelzpunkt: 168…171 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00151833
CAS-Nummer: 1546-80-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR