5-(2-Thienyl)pentanoic acid 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L13122.03
L13122.06
L13122.03EA
40.3
EUR
L13122.03
L13122.06
5-(2-Thienyl)pentanoic acid 95%
5-(2-Thienyl)pentanoic acid
Purity: 95%
CAS No.: 21010-06-0
Molecular Formula: C9H12O2S
Formula Weight: 184.26
Melting Point: 39-42°
Beilstein Reference: 126021
MDL No.: MFCD00151849
Formel:
C₉H₁₂O₂S Schmelzpunkt: 39…42 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00151849 CAS-Nummer: 21010-06-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...