5-(2-Thienyl)pentanoic acid 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 5-(Thiophen-2-yl)pentanoic acid

L13122.03 L13122.06
L13122.03EA 39.3 EUR
L13122.03 L13122.06
5-(2-Thienyl)pentanoic acid 95%
5-(2-Thienyl)pentanoic acid

Purity: 95% CAS No.: 21010-06-0 Molecular Formula: C9H12O2S Formula Weight: 184.26 Melting Point: 39-42° Beilstein Reference: 126021 MDL No.: MFCD00151849

Formel: C₉H₁₂O₂S
Molecular Weight: 184.26 g/mol
Schmelzpunkt: 39…42 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00151849
CAS-Nummer: 21010-06-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR