1-(Triisopropylsilyl)pyrrole 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L13152.06
L13152.14
L13152.06EA
78.6
EUR
L13152.06
L13152.14
1-(Triisopropylsilyl)pyrrole 95%
1-(Triisopropylsilyl)pyrrole
Purity: 95%
CAS No.: 87630-35-1
Molecular Formula: C13H25NSi
Formula Weight: 223.44
Flash Point: 106°(222°F)
Refractive Index: 1.4920
Boiling Point: 78°/0.4mm
Beilstein Reference: 3540663
MDL No.: MFCD00054932
Formel:
C₁₃H₂₅NSi Siedepunkt: 78 °C (0,4 mmHg) Dichte: 0,904 Flammpunkt: 106 °C (222 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00054932 CAS-Nummer: 87630-35-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...