Acridinorange (Zinkchlorid-Doppelsalz)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3,6-Bis(dimethylamino)acridine hydrochloride zinc chloride double salt, Euchrysine 3RX, Acridine orange hemi(Zinc chloride) salt, Acridine orange, Basic orange 14, 3,6-Bis(dimethylamino)acridinium chloride hemi(zinc chloride salt), Euchrysin GNY, N,N,N′,N′-Tetramethylacridine-3,6-diamine monohydrochloride compd. with zinc dichloride
L13159.06
L13159.14
L13159.06EA
23.5
EUR
L13159.06
L13159.14
Acridinorange (Zinkchlorid-Doppelsalz)
Acridinorange (Zinkchlorid-Doppelsalz)
CAS No.: 10127-02-3
Molecular Formula: C17H19N3.HCl.ZnCl2
Formula Weight: 438.10
Beilstein Reference: 3734978
MDL No.: MFCD00081043
Formel:
C₁₇H₂₀ClN₃·HCl·½ZnCl₂ Molecular Weight: 369.96 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00081043 CAS-Nummer: 10127-02-3 EINECS: 233-353-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...