Boran-Pyridin-Komplex 95%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
L13178.18
L13178.18EA
162
EUR
L13178.18
Boran-Pyridin-Komplex 95%
Boranepyridine complex
Purity: 95%
CAS No.: 110-51-0
Molecular Formula: C5H8BN
Formula Weight: 92.93
Storage Temperature: 2°C to 8°C
Flash Point: 73°(163°F)
Refractive Index: 1.5320
Odour: Unpleasant
Melting Point: 9-11°
Boiling Point: 65°/1mm
Beilstein Reference: 3674913
UN No.: UN2810
MDL No.: MFCD00012435
Formel:
C₅H₈BN Siedepunkt: 65 °C (1 mmHg) Schmelzpunkt: 9…11 °C Dichte: 0,920 Flammpunkt: 73 °C (163 °F) |
MDL:
MFCD00012435 CAS-Nummer: 110-51-0 EINECS: 203-773-4 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...