Mangan(IV)oxid tech. 90%, technical, activated

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Mangandioxid, Pyrolusit, Braunstein

L13253.22 L13253.09 L13253.36
L13253.22EA 31 EUR
L13253.22 L13253.09 L13253.36
Mangan(IV)oxid tech. 90%, technical, activated
Mangan(IV)oxid

Manganese(IV) oxide is suitable for use in batteries. It is widely used for the selective oxidation of allylic and benzylic alcohols to aldehydes or ketones, as a co-oxidant in combination with 1,4-benzoquinone and in the allylic oxidation of olefins catalyzed by Palladium(II)­ acetate. It acts a reagent for oxidations. It is the source of manganese and all its compounds, largely used in manufacturing of manganese steel, for making amethyst glass, decolorizing glass, painting on porcelain, faience and majolica. The precipitate is used in electrotechnics, pigments, browning gun barrels, drier for paints and varnishes, printing and dyeing textiles.

Formel: MnO₂
Molecular Weight: 86.94 g/mol
Schmelzpunkt: ∼ 535 °C (Zers.)
Dichte: 5.026
Lagertemperatur: Raumtemperatur
MDL: MFCD00003463
CAS-Nummer: 1313-13-9
Merck Index: 14,5730

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR