3,3',4,4'-Tetramethylbiphenyl 98%

Lieferant: Thermo Scientific

Synonyme: 4-(3,4-dimethylphenyl)-1,2-dimethylbenzene

L13338.03
L13338.03EA 51.2 EUR
L13338.03
3,3',4,4'-Tetramethylbiphenyl 98%
3,3',4,4'-Tetramethylbiphenyl

Purity: 98% CAS No.: 4920-95-0 Molecular Formula: C16H18 Formula Weight: 210.32 Melting Point: 74-77° Beilstein Reference: 1935370 MDL No.: MFCD00130219

Formel: C₁₆H₁₈
Molecular Weight: 210,32 g/mol
Schmelzpunkt: 74…77 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00130219
CAS-Nummer: 4920-95-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR