1-Boc-piperazin ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
tert-Butyl 1-piperazinecarboxylate, tert-butyl piperazine-1-carboxylate
L13363.06
L13363.14
L13363.03
L13363.06EA
124
EUR
L13363.06
L13363.14
L13363.03
1-Boc-piperazin ≥99%
1-Boc-piperazin
Purity: 99%
CAS No.: 57260-71-6
Molecular Formula: C9H18N2O2
Formula Weight: 186.26
Melting Point: 44-48°
Beilstein Reference: 879985
Formel:
C₉H₁₈N₂O₂ Schmelzpunkt: 44…48 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00075265 CAS-Nummer: 57260-71-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...