1,1,3,3-Tetramethyl-2-thioharnstoff ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Tetramethylthioharnstoff, Tetramethyl-2-thioharnstoff, N,N,N',N'-Tetramethylthioharnstoff

L13392.06 L13392.14
L13392.06EA 23.2 EUR
L13392.06 L13392.14
1,1,3,3-Tetramethyl-2-thioharnstoff ≥98%
1,1,3,3-Tetramethyl-2-thioharnstoff

Purity: 98% CAS No.: 2782-91-4 Molecular Formula: C5H12N2S Formula Weight: 132.23 Flash Point: 190°(374°F) Odour: Odourless Boiling Point: 245° Beilstein Reference: 1744916 MDL No.: MFCD00008324

Formel: C₅H₁₂N₂S
Molecular Weight: 132.23 g/mol
Siedepunkt: 245 °C
Schmelzpunkt: 75…77 °C
Flammpunkt: 190 °C (374 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008324
CAS-Nummer: 2782-91-4
EINECS: 220-488-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR