1-Pyrrolidinecarbonyl chloride 97%

Lieferant: Thermo Fisher Scientific
Gefahr

L13396.03 L13396.06
L13396.03EA 35.6 EUR
L13396.03 L13396.06
1-Pyrrolidinecarbonyl chloride 97%

Purity: 97% CAS No.: 1192-63-8 Molecular Formula: C5H8ClNO Formula Weight: 133.58 Storage Temperature: 2°C to 8°C Flash Point: 97°(206°F) Refractive Index: 1.4940 Boiling Point: 75°/1mm Beilstein Reference: 110281 UN No.: UN3265 MDL No.: MFCD00051321

Formel: C₅H₈ClNO
Siedepunkt: 75 °C (1 mmHg)
Dichte: 1,209
Flammpunkt: 97 °C (206 °F)
MDL: MFCD00051321
CAS-Nummer: 1192-63-8
EINECS: 214-758-7
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR