3,4,5-Trimethoxybenzylamin 96%
Lieferant: Thermo Fisher Scientific
L13461.06
L13461.14
L13461.06EA
62.8
EUR
L13461.06
L13461.14
3,4,5-Trimethoxybenzylamin 96%
Purity: 96%
CAS No.: 18638-99-8
Molecular Formula: C10H15NO3
Formula Weight: 197.24
Flash Point:
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...