N,N,N′,N′-Tetramethyl-O-(N-succinimidyl)-uroniumtetrafluoroborat 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: O-(N-Succinimidyl)-N,N,N',N'-tetramethyluronium Tetrafluoroborate, TSTU, 2-(2,5-Dioxopyrrolidin-1-yl)-1,1,3,3-tetramethylisouronium tetrafluoroborate

L13538.06 L13538.14
L13538.06EA 89.7 EUR
L13538.06 L13538.14
N,N,N′,N′-Tetramethyl-O-(N-succinimidyl)-uroniumtetrafluoroborat 98%
N,N,N′,N′-Tetramethyl-O-(N-succinimidyl)-uroniumtetrafluoroborat

Purity: 98% CAS No.: 105832-38-0 Molecular Formula: C9H16BF4N3O3 Formula Weight: 301.05 Melting Point: 195-200° MDL No.: MFCD00077875

Formel: C₉H₁₆BF₄N₃O₃
Molecular Weight: 301.05 g/mol
Schmelzpunkt: 195…200 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00077875
CAS-Nummer: 105832-38-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR