3,4-Dibrombenzaldehyd ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L13762.03
L13762.06
L13762.03EA
85.6
EUR
L13762.03
L13762.06
3,4-Dibrombenzaldehyd ≥99%
3,4-Dibromobenzaldehyde
Purity: 99%
CAS No.: 74003-55-7
Molecular Formula: C7H4Br2O
Formula Weight: 263.93
Melting Point: 69-72°
Beilstein Reference: 2573433
Formel:
C₇H₄Br₂O Schmelzpunkt: 69…72 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00236033 CAS-Nummer: 74003-55-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...