4-Brom-3-chlorphenol 98
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L13801.06
L13801.06EA
96.9
EUR
L13801.06
4-Brom-3-chlorphenol 98
4-Brom-3-chlorphenol
Purity: 98%
CAS No.: 13631-21-5
Molecular Formula: C6H4BrClO
Formula Weight: 207.46
Odour: Phenol-like
Melting Point: 64-68°
Beilstein Reference: 3236445
MDL No.: MFCD00070744
Formel:
C₆H₄BrClO Molecular Weight: 207.45 g/mol Schmelzpunkt: 64…68 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00070744 CAS-Nummer: 13631-21-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...