3-Chinolincarbaldehyd ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-Chinolincarboxaldehyd

L13815.03 L13815.06 L13815.MD
L13815.03EA 115 EUR
L13815.03 L13815.06 L13815.MD
3-Chinolincarbaldehyd ≥98%
3-Chinolincarbaldehyd

Purity: 98+% CAS No.: 13669-42-6 Molecular Formula: C10H7NO Formula Weight: 157.17 Melting Point: 68-71° Beilstein Reference: 113269

Formel: C₁₀H₇NO
Molecular Weight: 157.17 g/mol
Schmelzpunkt: 68…71 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006768
CAS-Nummer: 13669-42-6
EINECS: 237-147-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR