(S)-(+)-2-Dibenzylamino-3-phenyl-1-propanol 99%
Lieferant: Thermo Scientific
|
Achtung
|
L13916.06
L13916.06EA
34.6
EUR
L13916.06
(S)-(+)-2-Dibenzylamino-3-phenyl-1-propanol 99%
(S)-(+)-2-Dibenzylamino-3-phenyl-1-propanol
Purity: 99%
CAS No.: 111060-52-7
Molecular Formula: C23H25NO
Formula Weight: 331.46
Melting Point: 72-74°
Beilstein Reference: 7253058
Optical Rotation: +8.5° (c=4 in methanol)
MDL No.: MFCD00191984
Formel:
C₂₃H₂₅NO Molecular Weight: 331,46 g/mol Schmelzpunkt: 72…74 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00191984 CAS-Nummer: 111060-52-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...