Hydrogenperoxid-Harnstoff ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Urea hydrogen peroxide

L13940.14 L13940.22 L13940.36
L13940.14EA 23.1 EUR
L13940.14 L13940.22 L13940.36
Hydrogenperoxid-Harnstoff ≥97%
Hydrogenperoxid-Harnstoff

Purity: 97% CAS No.: 124-43-6 Molecular Formula: CH4N2O.H2O2 Formula Weight: 94.07 Odour: Odourless Melting Point: ca 90° dec. Beilstein Reference: 3680414 Merck Reference: 14,1782 UN No.: 1511 MDL No.: MFCD00013119

Formel: CH₆N₂O₃
Schmelzpunkt: ∼ 90 °C (Zersetzung)
Lagertemperatur: Kühlschrank
MDL: MFCD00013119
CAS-Nummer: 124-43-6
EINECS: 204-701-4
UN: 1511
ADR: 5.1,III
Merck Index: 14,1782

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR