2,2-Bis(hydroxymethyl)propionsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

L14014.36 L14014.22
L14014.36EA 67.3 EUR
L14014.36 L14014.22
2,2-Bis(hydroxymethyl)propionsäure ≥98%
2,2-Bis(hydroxymethyl)propionsäure

Purity: 98+% CAS No.: 4767-03-7 Molecular Formula: C5H10O4 Formula Weight: 134.13 Melting Point: 187-191° Beilstein Reference: 1756385 Merck Reference: 14,3253 MDL No.: MFCD00004199

Formel: C₅H₁₀O₄
Schmelzpunkt: 187…191 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004199
CAS-Nummer: 4767-03-7
EINECS: 225-306-3
Merck Index: 14,3253

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR