Sodium-3-chloro-2-hydroxypropanesulphonate hemihydrate 98%

Lieferant: Thermo Fisher Scientific
Achtung

L14018.36 L14018.22
L14018.36EA 105 EUR
L14018.36 L14018.22
Sodium-3-chloro-2-hydroxypropanesulphonate hemihydrate 98%
Sodium-3-chloro-2-hydroxypropanesulphonate hemihydrate

Purity: 98% CAS No.: 143218-48-8 Molecular Formula: C3H6ClNaO4S.0.5H2O Formula Weight: 205.60 (196.59anhy) Melting Point: ca 256° dec. Beilstein Reference: 4164376 MDL No.: MFCD00149699

Formel: C₃H₆ClNaO₄S·½H₂O
Molecular Weight: 205.60 g/mol (196.59 anhy)
Schmelzpunkt: ∼ 256 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149699
CAS-Nummer: 143218-48-8
EINECS: 204-807-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR