tert-Butylmethylether ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 99%
CAS No.: 1634-04-4
Molecular Formula: (CH3)3COCH3
Formula Weight: 88.15
Flash Point: -25°(-13°F)
Refractive Index: 1.3690
Odour: Mild
Melting Point: -109°
Boiling Point: 55-56°
Beilstein Reference: 1730942
Merck Reference: 14,6032
UN No.: 2350
MDL No.: MFCD00008812
Formel:
(CH₃)₃COCH₃ Siedepunkt: 55…56 °C Schmelzpunkt: –109 °C Dichte: 0,742 Flammpunkt: –25 °C (–13 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008812 CAS-Nummer: 1634-04-4 EINECS: 216-653-1 UN: 2350 ADR: 3,II Merck Index: 14,6032 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...