Adenosin-5'-monophosphorsäure (AMP), (max. 6% H₂O) 99% (Trockengewicht)

Lieferant: Thermo Fisher Scientific

Synonyme: ((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl dihydrogen phosphate, 5'-Adenylic acid, Adenosine 5'-monophosphate, Adenosine monophosphate, 5'-AMP

L14051.03 L14051.06
L14051.03EA 19.9 EUR
L14051.03 L14051.06
Adenosin-5'-monophosphorsäure (AMP), (max. 6% H₂O) 99% (Trockengewicht)
Adenosin-5'-monophosphorsäure (AMP)

a useful ligand determinant that facilitate the binding of APS reductase inhibitors and activates adenosine receptor agonists.


5'-AMP is an activator of a class of protein kinases known as AMP-activated protein kinase, an activator of phosphorylase b in the polymerization reaction for the synthesis of branched polysaccharides, a reagent in the synthesis of adenosine-5?-phosphoimidazolide, an inhibitor of endogenous NMN adenylyltransferase (NMNAT) activity that converts NMN to NAD+.

Formel: C₁₀H₁₄N₅O₇P
Molecular Weight: 347.22 g/mol
Schmelzpunkt: ∼ 190 °C (Zers.)
Lagertemperatur: Kühlschrank
MDL: MFCD00149360
CAS-Nummer: 61-19-8
EINECS: 200-500-0
Merck Index: 14,158

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR