2-Amino-4-(4-chlorphenyl)thiazol 98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
4-(4-Chlorophenyl)thiazol-2-amine
L14067.03
L14067.06
L14067.03EA
25.7
EUR
L14067.03
L14067.06
2-Amino-4-(4-chlorphenyl)thiazol 98%
2-Amino-4-(4-chlorophenyl)thiazole
Purity: 98%
CAS No.: 2103-99-3
Molecular Formula: C9H7ClN2S
Formula Weight: 210.69
Melting Point: 168-170°
Beilstein Reference: 153889
Formel:
C₉H₇ClN₂S Molecular Weight: 210.69 g/mol Schmelzpunkt: 168…170 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00047058 CAS-Nummer: 2103-99-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...