Tri-O-acetyl-D-glucal 98%

Lieferant: Thermo Scientific

Synonyme: (-)-Tri-O-acetyl-D-glucal, 1,5-Anhydro-2-deoxy-D-arabino-hex-1-enitol 3,4,6-Tri-O-acetyl Ether, (2R,3S,4R)-2-(Acetoxymethyl)-3,4-dihydro-2H-pyran-3,4-diyl diacetate

L14103.14
L14103.14EA 81.4 EUR
L14103.14
Tri-O-acetyl-D-glucal 98%
Tri-O-acetyl-D-glucal

Purity: 98% CAS No.: 2873-29-2 Molecular Formula: C12H16O7 Formula Weight: 272.26 Melting Point: 52-55° Beilstein Reference: 90781 Optical Rotation: -25° (c=1 in methanol) MDL No.: MFCD00063253

Formel: C₁₂H₁₆O₇
Molecular Weight: 272,25 g/mol
Schmelzpunkt: 52…55 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00063253
CAS-Nummer: 2873-29-2
EINECS: 220-709-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR