Amberlite® XAD4 Ionenaustauscher
Lieferant: Thermo Fisher Scientific
L14142.22
L14142.36
L14142.22EA
37.3
EUR
L14142.22
L14142.36
Amberlite® XAD4 Ionenaustauscher
Amberlite® XAD4
CAS No.: 37380-42-0
MDL No.: MFCD00132704
Lagertemperatur:
Raumtemperatur |
MDL:
MFCD00132704 CAS-Nummer: 37380-42-0 Merck Index: 14,382 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...