2,6-Di-tert-butyl-4-methylpyridin ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 97%
CAS No.: 38222-83-2
Molecular Formula: C14H23N
Formula Weight: 205.35
Storage Temperature: 2°C to 8°C
Flash Point: 83°(181°F)
Refractive Index: 1.4765
Melting Point: 30-35°
Boiling Point: 232-234°
Beilstein Reference: 130503
MDL No.: MFCD00006305
Formel:
C₁₄H₂₃N Siedepunkt: 232…234 °C Schmelzpunkt: 30…35 °C Flammpunkt: 83 °C (181 °F) Lagertemperatur: Kühlschrank |
MDL:
MFCD00006305 CAS-Nummer: 38222-83-2 EINECS: 253-834-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...