(1S)-(-)-Camphansäurechlorid 95%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: (1S)-4,7,7-Trimethyl-3-oxo-2-oxabicyclo[2.2.1]heptane-1-carbonyl chloride, (-)-Camphanic acid chloride

L14148.06
L14148.06EA 114 EUR
L14148.06
(1S)-(-)-Camphansäurechlorid 95%
(1S)-(-)-Camphansäurechlorid

Purity: 95% CAS No.: 39637-74-6 Molecular Formula: C10H13ClO3 Formula Weight: 216.67 Storage Temperature: -30°C to -10°C Melting Point: 64-68° Beilstein Reference: 3590860 Optical Rotation: -18° (c=2 in carbon tetrachloride) UN No.: 3261 MDL No.: MFCD00135626

Formel: C₁₀H₁₃ClO₃
Molecular Weight: 216.67 g/mol
Schmelzpunkt: 64…68 °C
MDL: MFCD00135626
CAS-Nummer: 39637-74-6
EINECS: 254-552-4
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR