2'-Desoxycytidin Hydrochlorid ≥99%

Lieferant: Thermo Fisher Scientific

Synonyme: 4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one hydrochloride

L14153.03 L14153.MD
L14153.03EA 57.4 EUR
L14153.03 L14153.MD
2'-Desoxycytidin Hydrochlorid ≥99%
2'-Desoxycytidin Hydrochlorid

Purity: 99% CAS No.: 3992-42-5 Molecular Formula: C9H13N3O4.HCl Formula Weight: 263.68 Odour: Odourless Melting Point: ca 170° dec. Beilstein Reference: 3576124 Optical Rotation: +57° (c=1 in water) MDL No.: MFCD00012840

Formel: C₉H₁₄ClN₃O₄
Molecular Weight: 263.68 g/mol
Schmelzpunkt: ∼ 170 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00012840
CAS-Nummer: 3992-42-5
EINECS: 223-639-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR